Learn More
Medchemexpress LLC Thalidomide-1-Me-5-NH2 | 1468761-60-5 | >98.0% | 287.27 g/mol | C14H13N3O4 | 100 MG
SDP
Catalog No. 5000283967
Encompass_Preferred
Thalidomide-1-Me-5-NH2 is an aminated thalidomide analogue used as an E3 ligase ligand building block in the design of PROTAC molecules; supplied in small milligram pack sizes for research use.
  • Aminated thalidomide analogue suitable as an E3 ligase ligand.
  • Primary amine available for derivatization and linker attachment.
  • High purity suitable for research applications.
  • Supplied in milligram-scale pack sizes for synthetic use.
  • Confirmed identity with a CAS registry number for unambiguous reference.
Catalog No. 50-002-83967 Supplier Medchemexpress LLC Supplier No. HYW586821100MG
Thalidomide-1-Me-5-NH2 is an aminated thalidomide analogue used as an E3 ligase ligand building block in the design of PROTAC molecules; supplied in small milligram pack sizes for research use.
  • Aminated thalidomide analogue suitable as an E3 ligase ligand.
  • Primary amine available for derivatization and linker attachment.
  • High purity suitable for research applications.
  • Supplied in milligram-scale pack sizes for synthetic use.
  • Confirmed identity with a CAS registry number for unambiguous reference.