Learn More
Medchemexpress LLC Thalidomide-1-Me-5-NH2 | 1468761-60-5 | >98.0% | 287.27 g/mol | C14H13N3O4 | 25 MG
SDP
Supplier:  Medchemexpress LLC HYW58682125MG
Encompass_Preferred
Thalidomide-1-Me-5-NH2 is an aminated analogue of thalidomide supplied as a research reagent. It acts as an E3 ligase ligand building block and can be derivatized at its amine for use in PROTAC design; it is not for medical, clinical diagnostic, or food use.
  • Aminated thalidomide analogue suitable as an E3 ligase ligand.
  • Can be derivatized at the amine for PROTAC and degrader design.
  • High purity (>98.0%) appropriate for chemical biology research.
  • Molecular formula C14H13N3O4, molecular weight 287.27 g/mol.
  • Available in small quantities suitable for synthesis, for example 25 MG sizes.
  • Supplied for research use only; not for medical or diagnostic use.
Catalog No. 50-002-83984