Learn More
Medchemexpress LLC Thalidomide 10 mM * 1 mL in DMSO Solution
SDP
Catalog No. NC3451858
Encompass_Preferred
Thalidomide inhibits cereblon (CRBN), a part of the cullin-4 E3 ubiquitin ligase complex CUL4-RBX1-DDB1, with a Kd of ∼250 nM, and has immunomodulatory, anti-inflammatory and anti-angiogenic cancer properties. Thalidomide can work as molecular glue to potentiate substrate.
Catalog No. NC3451858 Supplier Medchemexpress LLC Supplier No. HY1465810MM1MLDMSO
Thalidomide inhibits cereblon (CRBN), a part of the cullin-4 E3 ubiquitin ligase complex CUL4-RBX1-DDB1, with a Kd of ∼250 nM, and has immunomodulatory, anti-inflammatory and anti-angiogenic cancer properties. Thalidomide can work as molecular glue to potentiate substrate.