Learn More
Selleck Chemical LLC Thalidomide S1193-1g
SDP
Catalog No. 503110668
Encompass_Preferred
Thalidomide was introduced as a sedative drug immunomodulatory agent and also is investigated for treating symptoms of many cancers Thalidomide inhibits cereblon (CRBN) a part of the cullin-4 E3 ubiquitin ligase complex CUL4-RBX1-DDB1
Catalog No. 50-311-0668 Supplier Selleck Chemical LLC Supplier No. S11931G
Thalidomide was introduced as a sedative drug immunomodulatory agent and also is investigated for treating symptoms of many cancers Thalidomide inhibits cereblon (CRBN) a part of the cullin-4 E3 ubiquitin ligase complex CUL4-RBX1-DDB1