Learn More
Cayman Chemical -ThalIdomIde 1g
SDP
Supplier:  Cayman Chemical 146101 G
Encompass_Preferred
An immunomodulatory compound with diverse biological activities; prevents PMN chemotaxis at 1, 10, and 100 UG/ml; increases IL-2-induced proliferation and IFN-γ production in primary human T cells in vitro; enhances NK cell-mediated cytotoxicity in MM.1S multiple myeloma cells; reduces lung IL-6, TGF-β, VEGF, angiopoietin-1, angiopoietin-2, and collagen type Iα1 expression, inhibits pulmonary angiogenesis, and attenuates fibrosis in a mouse model of bleomycin-induced pulmonary fibrosis at 4 mg/animal; induces apoptosis in primary human embryonic fibroblasts (EC50 = 8.9 µM) and induces limb and eye defects in chicken embryos (EC50 = 50 UG/kg egg weight)
Catalog No. 50-297-2909