Learn More
Selleck Chemical LLC Thalidomide 200mg 50-35-1
SDP
Supplier:  Selleck Chemical LLC S1193200MG
Encompass_Preferred
Thalidomide was introduced as a sedative drug, immunomodulatory agent and also is investigated for treating symptoms of many cancers. Thalidomide inhibits an E3 ubiquitin ligase, which is a CRBN-DDB1-Cul4A complex. *For Research & Development use only.  Product is not intended for drug, household, or human consumption.
Catalog No. NC0433067