Learn More
Medchemexpress LLC Thalidomide-4,5-f | 2222115-19-5 | 98.0% | 294.21 g·mol⁻1 | C13H8F2N2O4 | 25 MG
SDP
Catalog No. 5000284907
Encompass_Preferred
Thalidomide-4,5-F is a fluorinated analogue of thalidomide used as a cereblon (E3 ligase) ligand in research. It is intended for use in PROTAC design and structure-activity studies, and the fluorine substituents allow further chemical derivatization.
  • Fluorinated thalidomide analogue that binds cereblon.
  • Suitable as an E3 ligase ligand for PROTAC research.
  • Enables further derivatization at fluorine positions.
  • High purity (≈98.0%) solid, white to off-white.
  • Molecular formula C13H8F2N2O4; molecular weight 294.21 g·mol⁻1.
  • Available in small research quantities, including 25 mg.
  • Store powder at -20 °C for long-term storage or 4 °C for short-term storage.
Catalog No. 50-002-84907 Supplier Medchemexpress LLC Supplier No. HYW57686825MG
Thalidomide-4,5-F is a fluorinated analogue of thalidomide used as a cereblon (E3 ligase) ligand in research. It is intended for use in PROTAC design and structure-activity studies, and the fluorine substituents allow further chemical derivatization.
  • Fluorinated thalidomide analogue that binds cereblon.
  • Suitable as an E3 ligase ligand for PROTAC research.
  • Enables further derivatization at fluorine positions.
  • High purity (≈98.0%) solid, white to off-white.
  • Molecular formula C13H8F2N2O4; molecular weight 294.21 g·mol⁻1.
  • Available in small research quantities, including 25 mg.
  • Store powder at -20 °C for long-term storage or 4 °C for short-term storage.