Learn More
Medchemexpress LLC Thalidomide-4,5-F | 2222115-19-5 | 98.0% | 294.21 g·mol⁻¹ | C13H8F2N2O4 | 100 MG
SDP
Catalog No. 5000284133
Encompass_Preferred
Thalidomide-4,5-F is a fluorinated analogue of thalidomide supplied for laboratory research. It promotes protein ubiquitinylation and serves as a cereblon-binding ligand; the fluorine substitutions allow further chemical derivatization. Intended for scientific research use only.
  • Fluorinated thalidomide analogue enabling further derivatization.
  • Promotes ubiquitinylation and binds cereblon for E3 ligase recruitment.
  • High purity (98.0%) suitable for research applications.
  • Available in multiple package sizes from 25 mg to 10 g.
  • White to off-white solid, suitable for storage and handling.
  • Recommended storage: powder at -20°C (up to 3 years) or 4°C (up to 2 years).
Catalog No. 50-002-84133 Supplier Medchemexpress LLC Supplier No. HYW576868100MG
Thalidomide-4,5-F is a fluorinated analogue of thalidomide supplied for laboratory research. It promotes protein ubiquitinylation and serves as a cereblon-binding ligand; the fluorine substitutions allow further chemical derivatization. Intended for scientific research use only.
  • Fluorinated thalidomide analogue enabling further derivatization.
  • Promotes ubiquitinylation and binds cereblon for E3 ligase recruitment.
  • High purity (98.0%) suitable for research applications.
  • Available in multiple package sizes from 25 mg to 10 g.
  • White to off-white solid, suitable for storage and handling.
  • Recommended storage: powder at -20°C (up to 3 years) or 4°C (up to 2 years).