Learn More
Medchemexpress LLC Thalidomide-4,5-F | 2222115-19-5 | 98.0% | 294.21 g/mol | C13H8F2N2O4 | 250 MG
SDP
Catalog No. 5000284540
Encompass_Preferred
Thalidomide-4,5-F is a fluorinated thalidomide analogue used as a cereblon (CRBN) ligand and E3 ligase recruiter for chemical biology and PROTAC research. It promotes ubiquitinylation of target proteins and can be derivatized at fluorine positions. Supplied as a solid powder with high purity and recommended cold storage conditions.
  • Fluorinated analogue designed for cereblon engagement.
  • High purity suitable for research applications.
  • Available in multiple milligram-scale pack sizes.
  • Stable solid form with defined cold storage conditions.
  • Modifiable at fluorine positions for derivatization.
Catalog No. 50-002-84540 Supplier Medchemexpress LLC Supplier No. HYW576868250MG
Thalidomide-4,5-F is a fluorinated thalidomide analogue used as a cereblon (CRBN) ligand and E3 ligase recruiter for chemical biology and PROTAC research. It promotes ubiquitinylation of target proteins and can be derivatized at fluorine positions. Supplied as a solid powder with high purity and recommended cold storage conditions.
  • Fluorinated analogue designed for cereblon engagement.
  • High purity suitable for research applications.
  • Available in multiple milligram-scale pack sizes.
  • Stable solid form with defined cold storage conditions.
  • Modifiable at fluorine positions for derivatization.