Learn More
Medchemexpress LLC Thalidomide-4,5-F | 2222115-19-5 | 98.0% | 294.21 g/mol | C13H8F2N2O4 | 500 MG
SDP
Catalog No. 5000284677
Encompass_Preferred
Thalidomide-4,5-F is a fluorinated analogue of thalidomide that functions as a cereblon (CRBN) E3 ligase ligand for chemical biology and targeted protein degradation research. Identifiers: CAS 2222115-19-5; formula C13H8F2N2O4; molecular weight 294.21 g/mol. It is supplied as a research-grade solid with reported purity around 98.0%.
  • Fluorinated analogue used as a cereblon E3 ligase ligand.
  • Enables recruitment of CRBN for targeted protein degradation studies.
  • Amenable to further derivatization via substitution at fluorine positions.
  • High chemical purity (approximately 98.0%).
  • Provided as a solid powder for research and analytical applications.
Catalog No. 50-002-84677 Supplier Medchemexpress LLC Supplier No. HYW576868500MG
Thalidomide-4,5-F is a fluorinated analogue of thalidomide that functions as a cereblon (CRBN) E3 ligase ligand for chemical biology and targeted protein degradation research. Identifiers: CAS 2222115-19-5; formula C13H8F2N2O4; molecular weight 294.21 g/mol. It is supplied as a research-grade solid with reported purity around 98.0%.
  • Fluorinated analogue used as a cereblon E3 ligase ligand.
  • Enables recruitment of CRBN for targeted protein degradation studies.
  • Amenable to further derivatization via substitution at fluorine positions.
  • High chemical purity (approximately 98.0%).
  • Provided as a solid powder for research and analytical applications.