Learn More
Medchemexpress LLC Thalidomide-4-Br | 2093536-12-8 | MFCD31730866 | 96.7% | 337.13 g/mol | C13H9BrN2O4 | 5 MG
SDP
Supplier:  Medchemexpress LLC HYW0391165MG
Encompass_Preferred
Thalidomide-4-Br is a brominated derivative of thalidomide supplied as a solid research reagent. It is active against bromodomains and is used in preclinical research on inflammation, tumor biology, and neurology. The compound is provided in small milligram quantities for screening, biochemical assays, and exploratory studies.
  • Brominated thalidomide derivative with defined activity toward bromodomains.
  • High purity suitable for biochemical and cellular research.
  • Available in small-quantity packs for early-stage studies.
  • Stable solid form with recommended cold storage for long-term retention.
  • Documented chemical identifiers, including CAS and molecular formula, for traceability.
Catalog No. 50-002-99745