Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Medchemexpress LLC Thalidomide-4-Br | 2093536-12-8 | MFCD31730866 | 99.6% | 337.13 | C13H9BrN2O4 | 50 MG
SDP

Supplier:  Medchemexpress LLC HYW03911650MG

Encompass_Preferred

Thalidomide-4-Br is a brominated thalidomide derivative used as a cereblon/bromodomain ligand in preclinical research on inflammation, oncology, and neurology. It is supplied as a solid, off-white to gray powder with characterized identity and stability data.

  • High purity (reported ≈99.6%).
  • Well characterized: CAS 2093536-12-8; formula C13H9BrN2O4; molecular weight 337.13.
  • Available in multiple package sizes, including small (50 mg) and larger quantities.
  • Defined storage and stability: powder -20°C (3 years) or 4°C (2 years); in solvent -80°C (6 months) or -20°C (1 month).
  • Suitable for cereblon-targeting studies and use as a ligand in PROTAC development.

Catalog No. 50-003-00531


May include imposed supplier surcharges.
Add to Cart

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.