Learn More
Medchemexpress LLC Thalidomide-4-Br | 2093536-12-8 | MFCD31730866 | 99.6% | 337.13 g·mol⁻¹ | C13H9BrN2O4 | 200 MG
SDP
Catalog No. 5000301910
Encompass_Preferred
Thalidomide-4-Br is a brominated thalidomide derivative used as a research compound that engages bromodomains and cereblon-related E3 ligase biology. It is applied in studies of inflammation, tumor biology, and neurology and is supplied as a high-purity solid with recommended cold storage for powder and solutions.
  • Engages bromodomains and E3 ligase pathways.
  • Used in inflammation, tumor, and neurology research.
  • Reported purity approximately 99.6% (COA/product listings).
  • Solid, off-white to gray physical form.
  • Storage: powder at -20°C (long term) or 4°C (short term); solutions at -80°C or -20°C.
  • Available in small-scale amounts and DMSO solution formats.
Catalog No. 50-003-01910 Supplier Medchemexpress LLC Supplier No. HYW039116200MG
Thalidomide-4-Br is a brominated thalidomide derivative used as a research compound that engages bromodomains and cereblon-related E3 ligase biology. It is applied in studies of inflammation, tumor biology, and neurology and is supplied as a high-purity solid with recommended cold storage for powder and solutions.
  • Engages bromodomains and E3 ligase pathways.
  • Used in inflammation, tumor, and neurology research.
  • Reported purity approximately 99.6% (COA/product listings).
  • Solid, off-white to gray physical form.
  • Storage: powder at -20°C (long term) or 4°C (short term); solutions at -80°C or -20°C.
  • Available in small-scale amounts and DMSO solution formats.