Learn More
Medchemexpress LLC Thalidomide-4-Br | 2093536-12-8 | MFCD31730866 | 99.7% | 337.13 g·mol⁻¹ | C13H9BrN2O4 | 10 MG
SDP
Catalog No. 5000299757
Encompass_Preferred
Thalidomide-4-Br is a brominated derivative of thalidomide that acts as a small-molecule bromodomain probe. It is provided as a high-purity solid for in vitro and preclinical research in inflammation, oncology, and neurology.
  • Bromodomain-containing small molecule probe.
  • Suitable for inflammation, tumor, and neurology research.
  • High purity: 99.7%.
  • Molecular formula C13H9BrN2O4; molecular weight 337.13 g·mol⁻¹.
  • Available in multiple pack sizes including 10 mg.
  • Powder storage: -20°C (3 years) or 4°C (2 years).
Catalog No. 50-002-99757 Supplier Medchemexpress LLC Supplier No. HYW03911610MG
Thalidomide-4-Br is a brominated derivative of thalidomide that acts as a small-molecule bromodomain probe. It is provided as a high-purity solid for in vitro and preclinical research in inflammation, oncology, and neurology.
  • Bromodomain-containing small molecule probe.
  • Suitable for inflammation, tumor, and neurology research.
  • High purity: 99.7%.
  • Molecular formula C13H9BrN2O4; molecular weight 337.13 g·mol⁻¹.
  • Available in multiple pack sizes including 10 mg.
  • Powder storage: -20°C (3 years) or 4°C (2 years).