Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Medchemexpress LLC Thalidomide 4-chloro | 244057-36-1 | MFCD30188071 | 96.0% | 292.68 g·mol⁻¹ | C13H9ClN2O4 | 1 G
SDP

Catalog No. 5000281481
Encompass_Preferred

Thalidomide 4-chloro is a chlorinated thalidomide derivative that functions as a cereblon (E3 ligase) ligand and E3 ligase activator. It is used in PROTAC development and as a synthetic building block for further derivatization via substitution at the 4-chloro position. The compound is supplied as a solid and is characterized for research applications.

  • Molecular formula: C13H9ClN2O4.
  • Molecular weight: 292.68 g·mol⁻¹.
  • CAS number: 244057-36-1.
  • Appearance: solid (powder).
  • Purity: 96.0%.
  • Intended use: E3 ligase (cereblon) ligand for PROTAC design and derivatization.
  • Storage: powder -20 °C (3 years) or 4 °C (2 years); in solvent -80 °C (6 months) or -20 °C (1 month).

Catalog No. 50-002-81481 Supplier Medchemexpress LLC Supplier No. HYW4601411G
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Thalidomide 4-chloro is a chlorinated thalidomide derivative that functions as a cereblon (E3 ligase) ligand and E3 ligase activator. It is used in PROTAC development and as a synthetic building block for further derivatization via substitution at the 4-chloro position. The compound is supplied as a solid and is characterized for research applications.

  • Molecular formula: C13H9ClN2O4.
  • Molecular weight: 292.68 g·mol⁻¹.
  • CAS number: 244057-36-1.
  • Appearance: solid (powder).
  • Purity: 96.0%.
  • Intended use: E3 ligase (cereblon) ligand for PROTAC design and derivatization.
  • Storage: powder -20 °C (3 years) or 4 °C (2 years); in solvent -80 °C (6 months) or -20 °C (1 month).

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.