Learn More
Medchemexpress LLC Thalidomide 4-chloro | 244057-36-1 | MFCD30188071 | >98.0% | 292.68 | C13H9ClN2O4 | 5 G
SDP
Supplier:  Medchemexpress LLC HYW4601415G
Encompass_Preferred
Thalidomide 4-chloro is a chlorinated thalidomide derivative that functions as an E3 ligase (cereblon) ligand/activator for research use. It is commonly used as a synthetic building block in PROTAC design and for studies involving CRBN recruitment, where the 4-chloro group can be substituted to create derivatives.
  • Functions as an E3 ligase (cereblon) ligand for biochemical studies.
  • Useful for PROTAC design and cereblon recruitment assays.
  • Enables further derivatization via substitution at the 4-chloro position.
  • Available in laboratory pack sizes suitable for synthesis and screening.
  • Analytical data and a product datasheet are available for reference.
Catalog No. 50-002-81480