Learn More
Medchemexpress LLC Thalidomide-4-F,5-Br | 2740657-28-5 | 99.4% | 355.12 g·mol⁻¹ | C13H8BrFN2O4 | 50 MG
SDP
Catalog No. 5000281732
Encompass_Preferred
Thalidomide-4-F,5-Br is a thalidomide analogue used as a cereblon (CRBN) E3 ligase ligand in biochemical research. It is provided as a purified powder for use in degradation studies, PROTAC design, and related cell biology and biochemical assays.
  • Acts as a cereblon E3 ligase ligand for targeted protein ubiquitination and degradation.
  • Suitable for PROTAC synthesis and biochemical assay development.
  • Supplied as a powder form for convenient handling and storage.
  • High purity (~99%) suitable for research applications.
  • Molecular weight approximately 355.12 g·mol⁻¹.
  • Available in multiple small package sizes for laboratory use.
Catalog No. 50-002-81732 Supplier Medchemexpress LLC Supplier No. HYW45371550MG
Thalidomide-4-F,5-Br is a thalidomide analogue used as a cereblon (CRBN) E3 ligase ligand in biochemical research. It is provided as a purified powder for use in degradation studies, PROTAC design, and related cell biology and biochemical assays.
  • Acts as a cereblon E3 ligase ligand for targeted protein ubiquitination and degradation.
  • Suitable for PROTAC synthesis and biochemical assay development.
  • Supplied as a powder form for convenient handling and storage.
  • High purity (~99%) suitable for research applications.
  • Molecular weight approximately 355.12 g·mol⁻¹.
  • Available in multiple small package sizes for laboratory use.