Learn More
Medchemexpress LLC Thalidomide-4-F,5-Br | 2740657-28-5 | MFCD34180776 | 99.4% | 355.12 g·mol⁻1 | C13H8BrFN2O4 | 25 MG
SDP
Catalog No. 5000281735
Encompass_Preferred
Thalidomide-4-F,5-Br is a halogenated thalidomide analogue that functions as an E3 ligase recruiting ligand for targeted protein degradation research. Supplied as a powder in small pre-weighed quantities, it is intended for research use in PROTAC development and studies of ubiquitin-mediated proteolysis.
  • Halogenated thalidomide derivative (fluoro and bromo substitutions).
  • Acts as a cereblon (CRBN) E3 ligase recruiting ligand for PROTAC applications.
  • High purity suitable for research (approximately 99.4%).
  • Provided as a powder for easy handling and formulation.
  • Available in small pre-weighed quantities for screening and synthesis workflows.
Catalog No. 50-002-81735 Supplier Medchemexpress LLC Supplier No. HYW45371525MG
Thalidomide-4-F,5-Br is a halogenated thalidomide analogue that functions as an E3 ligase recruiting ligand for targeted protein degradation research. Supplied as a powder in small pre-weighed quantities, it is intended for research use in PROTAC development and studies of ubiquitin-mediated proteolysis.
  • Halogenated thalidomide derivative (fluoro and bromo substitutions).
  • Acts as a cereblon (CRBN) E3 ligase recruiting ligand for PROTAC applications.
  • High purity suitable for research (approximately 99.4%).
  • Provided as a powder for easy handling and formulation.
  • Available in small pre-weighed quantities for screening and synthesis workflows.