Learn More
Medchemexpress LLC Thalidomide 4-fluoride | 835616-60-9 | 99.8% | 276.22 | 1 G
SDP
Catalog No. 5000404532
Encompass_Preferred
Thalidomide 4-fluoride (Cereblon ligand 4) is a Thalidomide-based Cereblon ligand utilized in the recruitment of the CRBN protein. It can be linked to the ligand for IRAK4 protein by a linker to form PROTAC IRAK4 degrader-1. Thalidomide targets cereblon (CRBN), a protein that is part of the cullin-4 E3 ubiquitin ligase complex, CUL4-RBX1-DDB1 (CRL4), which mediates the ubiquitination and subsequent proteasomal degradation of target proteins. The product appears as an off-white to gray solid.
  • Thalidomide-based Cereblon ligand
  • Used in the recruitment of CRBN protein
  • Can be connected to the ligand for IRAK4 protein to form PROTAC IRAK4 degrader-1
  • Targets cereblon (CRBN), part of the CUL4-RBX1-DDB1 E3 ubiquitin ligase complex
  • Mediates ubiquitination and proteasomal degradation of target proteins
Catalog No. 50-004-04532 Supplier Medchemexpress LLC Supplier No. HY415471G
Thalidomide 4-fluoride (Cereblon ligand 4) is a Thalidomide-based Cereblon ligand utilized in the recruitment of the CRBN protein. It can be linked to the ligand for IRAK4 protein by a linker to form PROTAC IRAK4 degrader-1. Thalidomide targets cereblon (CRBN), a protein that is part of the cullin-4 E3 ubiquitin ligase complex, CUL4-RBX1-DDB1 (CRL4), which mediates the ubiquitination and subsequent proteasomal degradation of target proteins. The product appears as an off-white to gray solid.
  • Thalidomide-based Cereblon ligand
  • Used in the recruitment of CRBN protein
  • Can be connected to the ligand for IRAK4 protein to form PROTAC IRAK4 degrader-1
  • Targets cereblon (CRBN), part of the CUL4-RBX1-DDB1 E3 ubiquitin ligase complex
  • Mediates ubiquitination and proteasomal degradation of target proteins