Learn More
Medchemexpress LLC Thalidomide 4-fluoride | 835616-60-9 | 99.8% | 276.22 | 10 G
SDP
Catalog No. 5000404956
Encompass_Preferred
Thalidomide 4-fluoride (Cereblon ligand 4) is a Cereblon-based ligand derived from Thalidomide. It functions by recruiting the CRBN protein. This compound can be linked to an IRAK4 protein ligand using a linker to create PROTAC IRAK4 degrader-1 (HY-129966).
  • Thalidomide-based Cereblon ligand
  • Used in the recruitment of CRBN protein
  • Can be connected to other ligands via a linker to form PROTACs
Catalog No. 50-004-04956 Supplier Medchemexpress LLC Supplier No. HY4154710G
Thalidomide 4-fluoride (Cereblon ligand 4) is a Cereblon-based ligand derived from Thalidomide. It functions by recruiting the CRBN protein. This compound can be linked to an IRAK4 protein ligand using a linker to create PROTAC IRAK4 degrader-1 (HY-129966).
  • Thalidomide-based Cereblon ligand
  • Used in the recruitment of CRBN protein
  • Can be connected to other ligands via a linker to form PROTACs