Learn More
Medchemexpress LLC Thalidomide 4-fluoride | 835616-60-9 | 99.8% | 276.22 | 100 G
SDP
Catalog No. 5000403627
Encompass_Preferred
Thalidomide 4-fluoride, also known as Cereblon ligand 4, is a Thalidomide-based Cereblon ligand. It is utilized in the recruitment of CRBN protein, a component of the cullin-4 E3 ubiquitin ligase complex. This compound mediates the ubiquitination and subsequent proteasomal degradation of target proteins, and can be connected to other ligands to form PROTAC degraders.
  • Thalidomide-based cereblon ligand.
  • Used in the recruitment of CRBN protein.
  • Targets cereblon (CRBN), a part of the E3 ubiquitin ligase complex.
  • Mediates ubiquitination and proteasomal degradation of target proteins.
  • Can be used to form PROTAC degraders.
Catalog No. 50-004-03627 Supplier Medchemexpress LLC Supplier No. HY41547100G
Thalidomide 4-fluoride, also known as Cereblon ligand 4, is a Thalidomide-based Cereblon ligand. It is utilized in the recruitment of CRBN protein, a component of the cullin-4 E3 ubiquitin ligase complex. This compound mediates the ubiquitination and subsequent proteasomal degradation of target proteins, and can be connected to other ligands to form PROTAC degraders.
  • Thalidomide-based cereblon ligand.
  • Used in the recruitment of CRBN protein.
  • Targets cereblon (CRBN), a part of the E3 ubiquitin ligase complex.
  • Mediates ubiquitination and proteasomal degradation of target proteins.
  • Can be used to form PROTAC degraders.