Learn More
Medchemexpress LLC Thalidomide 4-fluoride | 835616-60-9 | 99.9 % | 276.22 | 50 G
SDP
Supplier:  Medchemexpress LLC HY4154750G
Encompass_Preferred
Thalidomide 4-fluoride is a Thalidomide-based Cereblon ligand designed for the recruitment of CRBN protein. It can be linked to other ligands by a linker to form PROTAC degraders. In vitro, Thalidomide targets cereblon (CRBN), a ubiquitously expressed protein that is part of the cullin-4 E3 ubiquitin ligase complex. Cereblon mediates the ubiquitination and subsequent proteasomal degradation of target proteins.
  • Thalidomide-based cereblon ligand
  • Used in the recruitment of CRBN protein
  • Can be connected to other ligands to form PROTAC degraders
  • Targets cereblon (CRBN), a protein involved in the cullin-4 E3 ubiquitin ligase complex
  • Mediates ubiquitination and proteasomal degradation of target proteins
  • High purity for reliable research
  • Solid appearance, off-white to gray color
  • Stable under recommended storage conditions
  • Soluble in DMSO (200 mg/mL)
Catalog No. 50-004-04955