Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Medchemexpress LLC Thalidomide 4-fluoride (Cereblon ligand 4) | 835616-60-9 | 99.83% | 276.22 | 25 G
SDP

Catalog No. 5000402455
Encompass_Preferred

Thalidomide 4-fluoride (Cereblon ligand 4) is a Thalidomide-based Cereblon ligand that is utilized in the recruitment of the CRBN protein. It can be linked to the ligand for IRAK4 protein via a linker to form PROTAC IRAK4 degrader-1. It is for research use only and not sold to patients.

  • Purity: 99.83%
  • Molecular weight: 276.22
  • Formula: C13H9FN2O4
  • CAS No.: 835616-60-9
  • Appearance: Solid
  • Color: Off-white to gray
  • Solubility (in vitro): DMSO : 200 mg/mL (724.05 mM; requires ultrasonic; hygroscopic DMSO impacts solubility, use newly opened DMSO)
  • Storage (powder): -20°C for 3 years, 4°C for 2 years
  • Storage (in solvent): -80°C for 2 years, -20°C for 1 year

Catalog No. 50-004-02455 Supplier Medchemexpress LLC Supplier No. 25GHY41547
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Thalidomide 4-fluoride (Cereblon ligand 4) is a Thalidomide-based Cereblon ligand that is utilized in the recruitment of the CRBN protein. It can be linked to the ligand for IRAK4 protein via a linker to form PROTAC IRAK4 degrader-1. It is for research use only and not sold to patients.

  • Purity: 99.83%
  • Molecular weight: 276.22
  • Formula: C13H9FN2O4
  • CAS No.: 835616-60-9
  • Appearance: Solid
  • Color: Off-white to gray
  • Solubility (in vitro): DMSO : 200 mg/mL (724.05 mM; requires ultrasonic; hygroscopic DMSO impacts solubility, use newly opened DMSO)
  • Storage (powder): -20°C for 3 years, 4°C for 2 years
  • Storage (in solvent): -80°C for 2 years, -20°C for 1 year

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.