Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Medchemexpress LLC Thalidomide-4-O-C10-NH2 hydrochloride | 00-00-0 | 95.6% | C23H32ClN3O5 | 10MG
SDP

Catalog No. 5000261168
Encompass_Preferred

Thalidomide-4-O-C10-NH2 hydrochloride is a thalidomide-derived cereblon (CRBN) ligand formulated as a hydrochloride salt for use in PROTAC design and related biochemical research. Supplied as a solid with reported high purity, it is intended for small-scale laboratory studies and compound linking applications.

  • Hydrochloride salt form for improved stability
  • High purity suitable for research applications
  • Molecular formula C23H32ClN3O5 and molecular weight 465.97
  • Available in small packaging sizes for synthesis and screening
  • Solid form for convenient handling and storage

Catalog No. 50-002-61168 Supplier Medchemexpress LLC Supplier No. HY130963B10MG
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Thalidomide-4-O-C10-NH2 hydrochloride is a thalidomide-derived cereblon (CRBN) ligand formulated as a hydrochloride salt for use in PROTAC design and related biochemical research. Supplied as a solid with reported high purity, it is intended for small-scale laboratory studies and compound linking applications.

  • Hydrochloride salt form for improved stability
  • High purity suitable for research applications
  • Molecular formula C23H32ClN3O5 and molecular weight 465.97
  • Available in small packaging sizes for synthesis and screening
  • Solid form for convenient handling and storage

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.