Learn More
Medchemexpress LLC Thalidomide-4-o-c10-nh2 hydrochloride | 2460022-53-9 | 50 MG
SDP
Catalog No. 5000338554
Encompass_Preferred
Thalidomide-4-O-C10-NH2 hydrochloride is a thalidomide-based cereblon ligand used in the recruitment of CRBN protein. It is intended for use as a laboratory chemical and for the manufacture of substances, strictly for research purposes. This product has not been fully validated for medical applications.
  • Thalidomide-based cereblon ligand
  • Used in the recruitment of CRBN protein
  • For research use only
  • Intended for use as laboratory chemicals
  • Intended for the manufacture of substances
  • Recommended storage temperature: -20°C (solid)
  • For solutions, store at -80°C for 6 months or -20°C for 1 month
  • Shipped at room temperature if transit is less than 2 weeks
  • Store away from moisture and light
Catalog No. 50-003-38554 Supplier Medchemexpress LLC Supplier No. HY130963B50MG
Thalidomide-4-O-C10-NH2 hydrochloride is a thalidomide-based cereblon ligand used in the recruitment of CRBN protein. It is intended for use as a laboratory chemical and for the manufacture of substances, strictly for research purposes. This product has not been fully validated for medical applications.
  • Thalidomide-based cereblon ligand
  • Used in the recruitment of CRBN protein
  • For research use only
  • Intended for use as laboratory chemicals
  • Intended for the manufacture of substances
  • Recommended storage temperature: -20°C (solid)
  • For solutions, store at -80°C for 6 months or -20°C for 1 month
  • Shipped at room temperature if transit is less than 2 weeks
  • Store away from moisture and light