Learn More
Medchemexpress LLC Thalidomide-4-O-C10-NH2 hydrochloride | 95.6% | 465.97 | C23H32ClN3O5 | 5 MG
SDP
Supplier:  Medchemexpress LLC HY130963B5MG
Encompass_Preferred
Thalidomide-4-O-C10-NH2 hydrochloride is a thalidomide-derived cereblon (CRBN) ligand provided as a hydrochloride salt for use in PROTAC construction and chemical biology research. Supplied as a research reagent, it is intended for laboratory applications and not for human use.
  • Cereblon ligand suitable for E3 ligase recruitment in PROTACs and degrader design.
  • Hydrochloride salt improves solubility and handling compared with the free base.
  • Reported purity approximately 95.6% as supplied.
  • Molecular formula C23H32ClN3O5 and molecular weight 465.97.
  • Available in small research pack sizes, for example 5 mg.
Catalog No. 50-002-62931