Learn More
Medchemexpress LLC Thalidomide-4-O-C11-NH2 hydrochloride | 99.9% | 480.00 g·mol⁻¹ | C24H34ClN3O5 | 10 MG
SDP
Catalog No. 5000286657
Encompass_Preferred
Thalidomide-4-O-C11-NH2 hydrochloride is a thalidomide-derived cereblon (CRBN) ligand supplied as the hydrochloride salt for research use in targeted protein degradation and PROTAC development. It is intended as an E3 ligase ligand-linker conjugate for synthesis and biochemical studies involving CRBN recruitment.
  • High purity (99.94%) suitable for synthetic applications.
  • Hydrochloride salt form for improved handling and solubility.
  • Molecular formula C24H34ClN3O5; molecular weight 480.00 g·mol⁻¹.
  • Available in small mg-scale pack sizes for research workflows.
  • Designed for use as a cereblon ligand in PROTAC design and E3 ligase recruitment studies.
Catalog No. 50-002-86657 Supplier Medchemexpress LLC Supplier No. HY14957310MG
Thalidomide-4-O-C11-NH2 hydrochloride is a thalidomide-derived cereblon (CRBN) ligand supplied as the hydrochloride salt for research use in targeted protein degradation and PROTAC development. It is intended as an E3 ligase ligand-linker conjugate for synthesis and biochemical studies involving CRBN recruitment.
  • High purity (99.94%) suitable for synthetic applications.
  • Hydrochloride salt form for improved handling and solubility.
  • Molecular formula C24H34ClN3O5; molecular weight 480.00 g·mol⁻¹.
  • Available in small mg-scale pack sizes for research workflows.
  • Designed for use as a cereblon ligand in PROTAC design and E3 ligase recruitment studies.