Learn More
Medchemexpress LLC Thalidomide-4-O-C12-NH2 hydrochloride | 95.6% | 494.02 | C25H36ClN3O5 | 10 MG
SDP
Catalog No. 5000286812
Encompass_Preferred
Thalidomide-4-O-C12-NH2 hydrochloride is a thalidomide-derived cereblon ligand provided as the hydrochloride salt for use in chemical biology and medicinal chemistry applications. It functions as a CRBN E3 ligase recruiter for targeted protein degradation and is intended for use in PROTAC and ligand-linker conjugation studies. Store protected from light at recommended temperatures.
Catalog No. 50-002-86812 Supplier Medchemexpress LLC Supplier No. HY14957410MG
Thalidomide-4-O-C12-NH2 hydrochloride is a thalidomide-derived cereblon ligand provided as the hydrochloride salt for use in chemical biology and medicinal chemistry applications. It functions as a CRBN E3 ligase recruiter for targeted protein degradation and is intended for use in PROTAC and ligand-linker conjugation studies. Store protected from light at recommended temperatures.