Learn More
Medchemexpress LLC Thalidomide-4-O-C14-NH2 hydrochloride | 99.85% | 522.08 | 25 MG
SDP
Catalog No. 5000364041
Encompass_Preferred
Thalidomide-4-O-C14-NH2 hydrochloride is a Thalidomide (HY-10984)-based cereblon ligand that is used in the recruitment of CRBN protein. It can be linked to a protein ligand to create PROTACs. It is for research use only.
  • Functions as a ligand for E3 Ligase.
  • Involved in the recruitment of CRBN protein.
  • Can be connected via a linker to a protein ligand to form PROTACs.
  • Intended for research use only.
Catalog No. 50-003-64041 Supplier Medchemexpress LLC Supplier No. 25MGHY149576
Thalidomide-4-O-C14-NH2 hydrochloride is a Thalidomide (HY-10984)-based cereblon ligand that is used in the recruitment of CRBN protein. It can be linked to a protein ligand to create PROTACs. It is for research use only.
  • Functions as a ligand for E3 Ligase.
  • Involved in the recruitment of CRBN protein.
  • Can be connected via a linker to a protein ligand to form PROTACs.
  • Intended for research use only.