Learn More
Medchemexpress LLC Thalidomide-4-O-C14-NH2 hydrochloride | 99.9% | 522.08 | 5 MG
SDP
Supplier:  Medchemexpress LLC HY1495765MG
Encompass_Preferred
Thalidomide-4-O-C14-NH2 hydrochloride is a cereblon ligand derived from Thalidomide and is utilized for the recruitment of CRBN protein. It can be linked to a protein ligand via a linker to create PROTACs, which are relevant for research purposes.
  • Thalidomide-based cereblon ligand
  • Recruits CRBN protein
  • Can be connected to a protein ligand by a linker to form PROTACs
  • For research use only
Catalog No. 50-003-60019