Learn More
Medchemexpress LLC Thalidomide-4-O-C14-NH2 hydrochloride | 99.9% | 522.08 | C27H40ClN3O5 | 10 MG
SDP
Catalog No. 5000286813
Encompass_Preferred
Thalidomide-4-O-C14-NH2 hydrochloride is a carbon-14 radiolabeled thalidomide derivative used as a cereblon (CRBN) ligand in research applications, including PROTAC assembly. The product is supplied as the hydrochloride salt with high purity and characterized molecular properties for use in biochemical and pharmacokinetic studies.
Catalog No. 50-002-86813 Supplier Medchemexpress LLC Supplier No. HY14957610MG
Thalidomide-4-O-C14-NH2 hydrochloride is a carbon-14 radiolabeled thalidomide derivative used as a cereblon (CRBN) ligand in research applications, including PROTAC assembly. The product is supplied as the hydrochloride salt with high purity and characterized molecular properties for use in biochemical and pharmacokinetic studies.