Learn More
Medchemexpress LLC Thalidomide-4-O-C2-NH2 hydrochloride | 2341840-99-9 | 99.4% | 353.76 | 25 MG
SDP
Catalog No. 5000350189
Encompass_Preferred
Thalidomide-4-O-C2-NH2 hydrochloride is a synthesized E3 ligase ligand-linker conjugate. It incorporates a Thalidomide based cereblon ligand and a linker used in PROTAC technology, which exploits the intracellular ubiquitin-proteasome system to selectively degrade target proteins.
  • E3 ligase ligand-linker conjugate
  • Incorporates cereblon ligand
  • Utilizes PROTAC technology
  • Exploits ubiquitin-proteasome system
  • Selectively degrades target proteins
  • Targets Cereblon
  • Involved in Apoptosis and Autophagy pathways
Catalog No. 50-003-50189 Supplier Medchemexpress LLC Supplier No. 25MGHY136162
Thalidomide-4-O-C2-NH2 hydrochloride is a synthesized E3 ligase ligand-linker conjugate. It incorporates a Thalidomide based cereblon ligand and a linker used in PROTAC technology, which exploits the intracellular ubiquitin-proteasome system to selectively degrade target proteins.
  • E3 ligase ligand-linker conjugate
  • Incorporates cereblon ligand
  • Utilizes PROTAC technology
  • Exploits ubiquitin-proteasome system
  • Selectively degrades target proteins
  • Targets Cereblon
  • Involved in Apoptosis and Autophagy pathways