Learn More
Medchemexpress LLC Thalidomide-4-o-c2-nh2 hydrochloride | 2341840-99-9 | 99.4% | 353.76 | 50 MG
SDP
Supplier:  Medchemexpress LLC HY13616250MG
Encompass_Preferred
Thalidomide-4-O-C2-NH2 hydrochloride is a synthesized E3 ligase ligand-linker conjugate. It incorporates the thalidomide-based cereblon ligand and a linker used in PROTAC technology. This product is intended for research use only.
  • Synthesized E3 ligase ligand-linker conjugate
  • Incorporates thalidomide based cereblon ligand
  • Utilizes a linker used in PROTAC technology
  • For research use only
Catalog No. 50-003-50149