Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Medchemexpress LLC Thalidomide-4-O-C4-NH2 hydrochloride | 2376990-29-1 | MFCD32705123 | 99.4% | 381.81 | C17H20ClN3O5 | 5 MG
SDP

Catalog No. 5000262778
Encompass_Preferred

Thalidomide-4-O-C4-NH2 hydrochloride is a synthesized E3 ligase ligand-linker conjugate supplied as the hydrochloride salt for use in PROTAC research. It couples a thalidomide-derived cereblon ligand to a four-carbon alkyl linker terminating in a primary amine, enabling conjugation to target-binding moieties. The material is intended for laboratory research use only.

  • High purity (99.35%).
  • Hydrochloride salt form for handling and stability.
  • Molecular weight 381.81.
  • Molecular formula C17H20ClN3O5.
  • Available in small research quantities (5 mg and up).
  • Intended for use in PROTAC design and cereblon-based degrader development.

Catalog No. 50-002-62778 Supplier Medchemexpress LLC Supplier No. HY130948B5MG
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Thalidomide-4-O-C4-NH2 hydrochloride is a synthesized E3 ligase ligand-linker conjugate supplied as the hydrochloride salt for use in PROTAC research. It couples a thalidomide-derived cereblon ligand to a four-carbon alkyl linker terminating in a primary amine, enabling conjugation to target-binding moieties. The material is intended for laboratory research use only.

  • High purity (99.35%).
  • Hydrochloride salt form for handling and stability.
  • Molecular weight 381.81.
  • Molecular formula C17H20ClN3O5.
  • Available in small research quantities (5 mg and up).
  • Intended for use in PROTAC design and cereblon-based degrader development.

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.