Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Medchemexpress LLC Thalidomide-4-O-C4-NH2 hydrochloride | 2376990-29-1 | MFCD32705123 | 99.4% | C17H20ClN3O5 | 10MG
SDP

Catalog No. 5000261716
Encompass_Preferred

Thalidomide-4-O-C4-NH2 hydrochloride is a synthesized E3 ligase ligand-linker conjugate that combines a thalidomide-derived cereblon-binding moiety with a C4 amine-containing linker for use in PROTAC development and targeted protein degradation research. Supplied as a hydrochloride salt and intended for research use only.

  • Hydrochloride salt form for improved handling and solubility.
  • High purity suitable for biochemical and cell-based assays.
  • Molecular formula C17H20ClN3O5; molecular weight 381.81 g·mol⁻¹.
  • Available in small milligram quantities for discovery research.
  • Storage: sealed at 4°C; in solution, -80°C for long-term storage.

Catalog No. 50-002-61716 Supplier Medchemexpress LLC Supplier No. HY130948B10MG
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Thalidomide-4-O-C4-NH2 hydrochloride is a synthesized E3 ligase ligand-linker conjugate that combines a thalidomide-derived cereblon-binding moiety with a C4 amine-containing linker for use in PROTAC development and targeted protein degradation research. Supplied as a hydrochloride salt and intended for research use only.

  • Hydrochloride salt form for improved handling and solubility.
  • High purity suitable for biochemical and cell-based assays.
  • Molecular formula C17H20ClN3O5; molecular weight 381.81 g·mol⁻¹.
  • Available in small milligram quantities for discovery research.
  • Storage: sealed at 4°C; in solution, -80°C for long-term storage.

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.