Learn More
Medchemexpress LLC Thalidomide-4-O-C5-NH2 hydrochloride | 2419145-66-5 | 96.9% | 395.84 | 50 MG
Supplier: Medchemexpress LLC HY138848A50MG
Thalidomide-4-O-C5-NH2 hydrochloride is a synthesized E3 ligase ligand-linker conjugate, incorporating a Thalidomide-based cereblon ligand and a linker used in PROTAC technology. This compound exploits the intracellular ubiquitin-proteasome system to selectively degrade target proteins.
- E3 ligase ligand-linker conjugate
- Incorporates thalidomide-based cereblon ligand
- Used in PROTAC technology
- Targets E3 ligase ligand-linker conjugates
- Targets apoptosis and autophagy pathways
- Targets cereblon