Learn More
Medchemexpress LLC Thalidomide-4-O-C6-NH2 hydrochloride | 2245697-88-3 | 98.5% | 409.86 | C19H24ClN3O5 | 5 MG
SDP
Catalog No. 5000222250
Encompass_Preferred
Thalidomide-4-O-C6-NH2 hydrochloride is a synthesized E3 ligase ligand-linker conjugate supplied as the hydrochloride salt for research use. It is used as a building block in PROTAC dTAG-13, a degrader of FKBP12F36V and BET proteins, and is provided with supporting analytical data and handling documentation.
  • Provided as the hydrochloride salt suitable for synthetic applications.
  • High purity (98.5% by HPLC) with supporting analytical data.
  • Analytical files include data sheet, COA, H NMR, and LCMS.
  • Available in multiple pack sizes from milligrams to grams.
  • Intended for research use only; not for human or therapeutic use.
Catalog No. 50-002-22250 Supplier Medchemexpress LLC Supplier No. HY135250B5MG
Thalidomide-4-O-C6-NH2 hydrochloride is a synthesized E3 ligase ligand-linker conjugate supplied as the hydrochloride salt for research use. It is used as a building block in PROTAC dTAG-13, a degrader of FKBP12F36V and BET proteins, and is provided with supporting analytical data and handling documentation.
  • Provided as the hydrochloride salt suitable for synthetic applications.
  • High purity (98.5% by HPLC) with supporting analytical data.
  • Analytical files include data sheet, COA, H NMR, and LCMS.
  • Available in multiple pack sizes from milligrams to grams.
  • Intended for research use only; not for human or therapeutic use.