Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Medchemexpress LLC Thalidomide-4-O-C6-NH2 hydrochloride | 2245697-88-3 | 98.5% | C19H24ClN3O5 | 10MG
SDP

Catalog No. 5000258493
Encompass_Preferred

Thalidomide-4-O-C6-NH2 hydrochloride is a synthesized E3 ligase ligand-linker conjugate used in PROTAC research, notably as a component of the degrader dTAG-13 that directs FKBP12F36V and BET proteins for degradation. It is intended for laboratory research into E3 ligase recruitment, targeted protein degradation, apoptosis, and autophagy.

  • High purity suitable for research (98.5%).
  • Soluble in DMSO at 83.33 mg/mL (203.31 mM); ultrasonic assistance may be needed.
  • Engages cereblon for E3 ligase-mediated protein degradation.
  • Useful for PROTAC assembly and linker conjugation workflows.
  • Available in small research quantities to support assay development.
  • Recommended storage: 4°C sealed, away from moisture and light; in solvent, store at -80°C for long term.

Catalog No. 50-002-58493 Supplier Medchemexpress LLC Supplier No. HY135250B10MG
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Thalidomide-4-O-C6-NH2 hydrochloride is a synthesized E3 ligase ligand-linker conjugate used in PROTAC research, notably as a component of the degrader dTAG-13 that directs FKBP12F36V and BET proteins for degradation. It is intended for laboratory research into E3 ligase recruitment, targeted protein degradation, apoptosis, and autophagy.

  • High purity suitable for research (98.5%).
  • Soluble in DMSO at 83.33 mg/mL (203.31 mM); ultrasonic assistance may be needed.
  • Engages cereblon for E3 ligase-mediated protein degradation.
  • Useful for PROTAC assembly and linker conjugation workflows.
  • Available in small research quantities to support assay development.
  • Recommended storage: 4°C sealed, away from moisture and light; in solvent, store at -80°C for long term.

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.