Learn More
Medchemexpress LLC Thalidomide-4-O-C7-NH2 hydrochloride | 00-00-0 | 99.8% | C20H26ClN3O5 | 10MG
SDP
Supplier:  Medchemexpress LLC HY130949B10MG
Encompass_Preferred
Thalidomide-4-O-C7-NH2 hydrochloride is a thalidomide-derived cereblon (CRBN) ligand-linker conjugate provided as the hydrochloride salt for research use in PROTAC development and E3 ligase recruitment studies. It is supplied as a high-purity, well-characterized reagent suitable for biochemical assays, linker conjugation, and medicinal chemistry workflows.
  • High purity (99.8%) suitable for research applications.
  • Hydrochloride salt form for consistent handling and storage.
  • Characterized molecular formula C20H26ClN3O5 and molecular weight 387.43 g/mol.
  • Functions as a cereblon (CRBN) E3 ligase ligand for PROTAC design.
  • Available in small, lab-scale quantities for screening and synthesis.
  • Datasheet and certificate of analysis available from the supplier.
Catalog No. 50-002-61971