Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Medchemexpress LLC Thalidomide-4-O-C7-NH2 hydrochloride | 99.8% | 423.89 g/mol | C20H26ClN3O5 | 5 MG
SDP

Catalog No. 5000262160
Encompass_Preferred

Thalidomide-4-O-C7-NH2 hydrochloride is a thalidomide-derived cereblon ligand-linker conjugate used as an E3 ligase ligand in PROTAC development and CRBN recruitment studies.

  • High chemical purity: 99.8%.
  • Hydrochloride salt form for improved stability and solubility.
  • Molecular formula C20H26ClN3O5; molecular weight 423.89 g/mol.
  • Optimized for use as an E3 ligase ligand in biochemical and cell-based assays.
  • Supplied in small research quantities suitable for synthesis and assay development.

Catalog No. 50-002-62160 Supplier Medchemexpress LLC Supplier No. HY130949B5MG
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Thalidomide-4-O-C7-NH2 hydrochloride is a thalidomide-derived cereblon ligand-linker conjugate used as an E3 ligase ligand in PROTAC development and CRBN recruitment studies.

  • High chemical purity: 99.8%.
  • Hydrochloride salt form for improved stability and solubility.
  • Molecular formula C20H26ClN3O5; molecular weight 423.89 g/mol.
  • Optimized for use as an E3 ligase ligand in biochemical and cell-based assays.
  • Supplied in small research quantities suitable for synthesis and assay development.

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.