Learn More
Medchemexpress LLC Thalidomide-4-O-C9-NH2 hydrochloride | >98.0% | 451.94 g/mol | C22H30ClN3O5 | 10 MG
SDP
Catalog No. 5000285802
Encompass_Preferred
Thalidomide-4-O-C9-NH2 hydrochloride is a thalidomide-derived cereblon (CRBN) ligand intended for use in targeted protein degradation and PROTAC development. Supplied as the hydrochloride salt for conjugation chemistry, the compound is offered for research use and is commonly provided in small milligram quantities for medicinal chemistry and chemical biology applications.
  • Acts as a cereblon (CRBN) ligand for E3 ligase recruitment.
  • Designed for use in PROTAC and targeted protein degradation research.
  • Provided as a hydrochloride salt suitable for conjugation chemistry.
  • Reported purity ≥98% according to vendor listings.
  • Molecular weight ~451.94 g/mol; formula C22H30ClN3O5.
  • Store sealed at -20°C to maintain stability.
Catalog No. 50-002-85802 Supplier Medchemexpress LLC Supplier No. HY14957210MG
Thalidomide-4-O-C9-NH2 hydrochloride is a thalidomide-derived cereblon (CRBN) ligand intended for use in targeted protein degradation and PROTAC development. Supplied as the hydrochloride salt for conjugation chemistry, the compound is offered for research use and is commonly provided in small milligram quantities for medicinal chemistry and chemical biology applications.
  • Acts as a cereblon (CRBN) ligand for E3 ligase recruitment.
  • Designed for use in PROTAC and targeted protein degradation research.
  • Provided as a hydrochloride salt suitable for conjugation chemistry.
  • Reported purity ≥98% according to vendor listings.
  • Molecular weight ~451.94 g/mol; formula C22H30ClN3O5.
  • Store sealed at -20°C to maintain stability.