Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Medchemexpress LLC Thalidomide-4-O-C9-NH2 (hydrochloride) | 99.3% | 451.94 | 25 MG
SDP

Catalog No. 5000363970
Encompass_Preferred

Thalidomide-4-O-C9-NH2 hydrochloride is a chemical compound for laboratory research and the manufacture of substances. This compound is stable under recommended storage conditions, which include sealed storage at -20°C, away from moisture and light. If in solvent, it can be stored at -80°C for 6 months or -20°C for 1 month.

  • Intended for laboratory research and substance manufacturing
  • Molecular weight of 451.94
  • Purity of 99.26%
  • Stable under recommended storage conditions
  • Store at -20°C, sealed, away from moisture and light
  • In solvent, store at -80°C for 6 months or -20°C for 1 month
  • Can be shipped at room temperature if transit is less than 2 weeks

Catalog No. 50-003-63970 Supplier Medchemexpress LLC Supplier No. 25MGHY149572
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Thalidomide-4-O-C9-NH2 hydrochloride is a chemical compound for laboratory research and the manufacture of substances. This compound is stable under recommended storage conditions, which include sealed storage at -20°C, away from moisture and light. If in solvent, it can be stored at -80°C for 6 months or -20°C for 1 month.

  • Intended for laboratory research and substance manufacturing
  • Molecular weight of 451.94
  • Purity of 99.26%
  • Stable under recommended storage conditions
  • Store at -20°C, sealed, away from moisture and light
  • In solvent, store at -80°C for 6 months or -20°C for 1 month
  • Can be shipped at room temperature if transit is less than 2 weeks

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.