Learn More
Medchemexpress LLC Thalidomide-4-O-CH2-COO(t-Bu) | 1950635-36-5 | 99.26% | 388.37 | 100 MG
SDP
Catalog No. 5000403576
Encompass_Preferred
Thalidomide-4-O-CH2-COO(t-Bu) is a t-Bu modified Thalidomide, which is the final step intermediate in the synthesis of Thalidomide-4-OH. It acts as a Cereblon ligand to recruit CRBN proteins. The t-Bu protecting group can be removed under acidic conditions to participate in the synthesis of PROTAC molecules. It is a key intermediate in the synthesis of CRBN-based designed PROTAC molecules.
  • Acts as a Cereblon ligand to recruit CRBN proteins.
  • The t-Bu protecting group can be removed under acidic conditions to participate in the synthesis of PROTAC molecules.
  • Key intermediate in the synthesis of CRBN-based designed PROTAC molecules.
Catalog No. 50-004-03576 Supplier Medchemexpress LLC Supplier No. HY42771100MG
Thalidomide-4-O-CH2-COO(t-Bu) is a t-Bu modified Thalidomide, which is the final step intermediate in the synthesis of Thalidomide-4-OH. It acts as a Cereblon ligand to recruit CRBN proteins. The t-Bu protecting group can be removed under acidic conditions to participate in the synthesis of PROTAC molecules. It is a key intermediate in the synthesis of CRBN-based designed PROTAC molecules.
  • Acts as a Cereblon ligand to recruit CRBN proteins.
  • The t-Bu protecting group can be removed under acidic conditions to participate in the synthesis of PROTAC molecules.
  • Key intermediate in the synthesis of CRBN-based designed PROTAC molecules.