Learn More
Medchemexpress LLC Thalidomide-4-O-CH2-COO(t-Bu) | 1950635-36-5 | MFCD32643096 | 99.3% | 388.37 | C19H20N2O7 | 10 MG
SDP
Catalog No. 5000299246
Encompass_Preferred
Thalidomide-4-O-CH2-COO(t-Bu) is a tert-butyl protected thalidomide derivative used as a cereblon (CRBN) binding ligand and as a synthetic intermediate in the preparation of PROTAC molecules. It is supplied for research use only and is not intended for human use.
  • Tert-butyl protected thalidomide derivative suitable for deprotection under acidic conditions.
  • Functions as a cereblon (CRBN) binding ligand for use in PROTAC design.
  • High reported purity (~99.3%) for reliable research applications.
  • Available in small research pack sizes (mg scale) for synthetic workflows.
  • Stable at room temperature for extended storage; follow solvent storage recommendations for longer-term stability.
Catalog No. 50-002-99246 Supplier Medchemexpress LLC Supplier No. HY4277110MG
Thalidomide-4-O-CH2-COO(t-Bu) is a tert-butyl protected thalidomide derivative used as a cereblon (CRBN) binding ligand and as a synthetic intermediate in the preparation of PROTAC molecules. It is supplied for research use only and is not intended for human use.
  • Tert-butyl protected thalidomide derivative suitable for deprotection under acidic conditions.
  • Functions as a cereblon (CRBN) binding ligand for use in PROTAC design.
  • High reported purity (~99.3%) for reliable research applications.
  • Available in small research pack sizes (mg scale) for synthetic workflows.
  • Stable at room temperature for extended storage; follow solvent storage recommendations for longer-term stability.