Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Medchemexpress LLC Thalidomide-4-OH | 5054-59-1 | MFCD03699892 | 99.9% | 274.23 g·mol⁻¹ | C13H10N2O5 | 10 G
SDP

Catalog No. 5000247645
Encompass_Preferred

Thalidomide-4-OH is a 4-hydroxylated metabolite of thalidomide used as a Cereblon (CRBN) ligand in chemical biology and PROTAC research. It is supplied as a high-purity solid for synthetic and biochemical applications, with recommended storage under inert atmosphere.

  • High purity suitable for synthesis and assays.
  • Molecular formula C13H10N2O5; molecular weight 274.23 g·mol⁻¹.
  • Used as a CRBN ligand for recruitment of cereblon in PROTACs.
  • Available in gram-scale quantities for research workflows.
  • Solid with melting point 273-275 °C.
  • Recommended storage: 4 °C under nitrogen; in solvent: -80 °C (6 months) or -20 °C (1 month).

Catalog No. 50-002-47645 Supplier Medchemexpress LLC Supplier No. HY10359610G
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Thalidomide-4-OH is a 4-hydroxylated metabolite of thalidomide used as a Cereblon (CRBN) ligand in chemical biology and PROTAC research. It is supplied as a high-purity solid for synthetic and biochemical applications, with recommended storage under inert atmosphere.

  • High purity suitable for synthesis and assays.
  • Molecular formula C13H10N2O5; molecular weight 274.23 g·mol⁻¹.
  • Used as a CRBN ligand for recruitment of cereblon in PROTACs.
  • Available in gram-scale quantities for research workflows.
  • Solid with melting point 273-275 °C.
  • Recommended storage: 4 °C under nitrogen; in solvent: -80 °C (6 months) or -20 °C (1 month).

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.