Learn More
Medchemexpress LLC Thalidomide-4-OH | 5054-59-1 | MFCD03699892 | 99.9% | 274.23 g/mol | C13H10N2O5 | 5 G
SDP
Catalog No. 5000245164
Encompass_Preferred
Thalidomide-4-OH is a 4-hydroxylated thalidomide derivative that serves as a cereblon (CRBN) ligand for targeted protein degradation research. It is commonly used as a building block in PROTAC design and medicinal chemistry studies and is supplied as a solid for laboratory research use only.
  • Acts as a cereblon (CRBN) ligand suitable for PROTAC synthesis.
  • 4-hydroxyl substitution enables linker attachment for conjugation.
  • High purity supports reproducible conjugation and biological assays.
  • Solid form allows convenient storage and handling.
  • Useful for ligand optimization and E3 ligase recruitment studies.
Catalog No. 50-002-45164 Supplier Medchemexpress LLC Supplier No. HY1035965G
Thalidomide-4-OH is a 4-hydroxylated thalidomide derivative that serves as a cereblon (CRBN) ligand for targeted protein degradation research. It is commonly used as a building block in PROTAC design and medicinal chemistry studies and is supplied as a solid for laboratory research use only.
  • Acts as a cereblon (CRBN) ligand suitable for PROTAC synthesis.
  • 4-hydroxyl substitution enables linker attachment for conjugation.
  • High purity supports reproducible conjugation and biological assays.
  • Solid form allows convenient storage and handling.
  • Useful for ligand optimization and E3 ligase recruitment studies.