Learn More
Medchemexpress LLC Thalidomide-5 6-F 1g
SDP
Supplier:  Medchemexpress LLC HYW0932721G
Encompass_Preferred
Thalidomide-5 6-F is the Thalidomide-based cereblon ligand used in the recruitment of CRBN protein Thalidomide-5 6-F can be connected to the ligand for protein by a linker to form PROTACs[1]
Catalog No. 50-002-04542