Learn More
Medchemexpress LLC Thalidomide-5-Br | 26166-92-7 | 98.4% | 337.13 g/mol | C13H9BrN2O4 | 50 MG
SDP
Catalog No. 5000210246
Encompass_Preferred
Thalidomide-5-Br is a brominated thalidomide analogue supplied for biochemical research. It has a reported molecular weight of 337.13 g/mol and is supplied as an off-white to gray solid with reported purity around 98%.
  • Used in research of inflammation, tumor biology, and neurology.
  • Supplied as a solid suitable for formulation or dissolution in solvent for experimental use.
  • Reported high purity for analytical and preclinical studies.
  • Stable at room temperature for extended storage; in-solution storage requires low temperatures.
Catalog No. 50-002-10246 Supplier Medchemexpress LLC Supplier No. HYW06960450MG
Thalidomide-5-Br is a brominated thalidomide analogue supplied for biochemical research. It has a reported molecular weight of 337.13 g/mol and is supplied as an off-white to gray solid with reported purity around 98%.
  • Used in research of inflammation, tumor biology, and neurology.
  • Supplied as a solid suitable for formulation or dissolution in solvent for experimental use.
  • Reported high purity for analytical and preclinical studies.
  • Stable at room temperature for extended storage; in-solution storage requires low temperatures.