Learn More
Medchemexpress LLC Thalidomide-5-CH2-NH2 hydrochloride | 1010100-22-7 | MFCD32666042 | 98.6% | 323.73 | C14H14ClN3O4 | 50 MG
SDP
Catalog No. 5000282794
Encompass_Preferred
Thalidomide-5-CH2-NH2 hydrochloride is a thalidomide-derived cereblon (CRBN) ligand used as an E3 ligase recruiting moiety in PROTAC design and targeted protein degradation research. Supplied as a solid with high purity, it serves as a linker-compatible building block for medicinal chemistry and chemical biology applications.
  • High purity suitable for research.
  • Solid form, easy to handle and store.
  • Compatible with linker conjugation for PROTAC synthesis.
  • Facilitates CRBN-mediated target recruitment.
  • Available in multiple small pack sizes for synthesis workflows.
Catalog No. 50-002-82794 Supplier Medchemexpress LLC Supplier No. HYW12987250MG
Thalidomide-5-CH2-NH2 hydrochloride is a thalidomide-derived cereblon (CRBN) ligand used as an E3 ligase recruiting moiety in PROTAC design and targeted protein degradation research. Supplied as a solid with high purity, it serves as a linker-compatible building block for medicinal chemistry and chemical biology applications.
  • High purity suitable for research.
  • Solid form, easy to handle and store.
  • Compatible with linker conjugation for PROTAC synthesis.
  • Facilitates CRBN-mediated target recruitment.
  • Available in multiple small pack sizes for synthesis workflows.