Learn More
Medchemexpress LLC Thalidomide-5-CH2-NH2 hydrochloride | 1010100-22-7 | MFCD32666042 | 98.6% | 323.73 g/mol | C14H14ClN3O4 | 25 MG
SDP
Supplier:  Medchemexpress LLC HYW12987225MG
Encompass_Preferred
Thalidomide-5-CH2-NH2 hydrochloride is a thalidomide-derived cereblon (CRBN) ligand supplied as a hydrochloride salt. It provides a reactive primary amine at the 5-position for linker attachment and is used as an E3 ligase recruiting moiety in PROTAC design and related chemical biology research. Suitable for medicinal chemistry and linker-conjugation workflows.
Catalog No. 50-002-83261